1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

PC8571-1G
APOSPC8571-1GEA 538 EUR
APOSPC8571-1G
1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Formel: C₁₃H₉ClFNO₂
Molecular Weight: 265,67 g/mol
Schmelzpunkt: 130…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817431
CAS-Nummer: 952183-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR