4-Fluor-N-methylbenzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
PC8577-1G
APOSPC8577-1GEA
0
EUR
APOSPC8577-1G
4-Fluor-N-methylbenzolsulfonamid
4-Fluor-N-methylbenzolsulfonamid
Formel:
C₇H₈FNO₂S Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01211989 CAS-Nummer: 433-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...