N-Methyl-3-(trifluoromethyl)benzenesulphonamide
Lieferant: Apollo Scientific
|
Achtung
|
PC8578-1G
APOSPC8578-1GEA
222
EUR
APOSPC8578-1G
N-Methyl-3-(trifluoromethyl)benzenesulphonamide
N-Methyl-3-(trifluoromethyl)benzenesulphonamide
Formel:
C₈H₈F₃NO₂S Schmelzpunkt: 62…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817446 CAS-Nummer: 882423-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...