2,3,4,5,6-Pentafluorobenzyl chloroformate 97%

Lieferant: Apollo Scientific
Gefahr

PC8579-1G
APOSPC8579-1GEA 207 EUR
APOSPC8579-1G
2,3,4,5,6-Pentafluorobenzyl chloroformate 97%
2,3,4,5,6-Pentafluorobenzyl chloroformate

Derivatisation agent for the analysis of amines, alcohols, and amino acids by Electron-Capture Negative Ion Chemical Ionisation-Mass Spectrometry.

Formel: C₈H₂ClF₅O₂
Molecular Weight: 260,55 g/mol
Siedepunkt: 116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00798136
CAS-Nummer: 53526-74-2
UN: 3277
ADR: 6.1+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR