6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

Synonyme: 1,6-Dihydro-6-oxo-1-[3-(trifluoromethyl)benzyl]pyridine-3-carboxaldehyde

PC8580-1G
APOSPC8580-1GEA 538 EUR
APOSPC8580-1G
6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridinecarbaldehyde
6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridinecarbaldehyde
Formel: C₁₄H₁₀F₃NO₂
Molecular Weight: 281,24 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817436
CAS-Nummer: 952183-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR