5-(Chloromethyl)-2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole

Lieferant: Apollo Scientific
Achtung

PC8583-500MG
APOSPC8583-500MGEA 332 EUR
APOSPC8583-500MG
5-(Chloromethyl)-2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole
5-(Chloromethyl)-2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole
Formel: C₁₂H₈ClF₄NS
Molecular Weight: 309,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07369415
CAS-Nummer: 317319-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR