2-[(4-Fluorobenzyl)sulphanyl]ethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC8586-1G PC8586-250MG
APOSPC8586-1GEA 205 EUR
APOSPC8586-1G APOSPC8586-250MG
2-[(4-Fluorobenzyl)sulphanyl]ethylamine hydrochloride
2-[(4-Fluorobenzyl)sulphanyl]ethylamine hydrochloride
Formel: C₉H₁₃ClFNS
Molecular Weight: 221.73 g/mol
Schmelzpunkt: 167…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027133
CAS-Nummer: 1170404-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR