2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid

Lieferant: Apollo Scientific

PC8590-500MG
APOSPC8590-500MGEA 438 EUR
APOSPC8590-500MG
2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Formel: C₁₁H₅F₄NO₂S
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027138
CAS-Nummer: 937602-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR