{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl}methanol

Lieferant: Apollo Scientific

Synonyme: 5-(Hydroxymethyl)-2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole

PC8593-500MG
APOSPC8593-500MGEA 268 EUR
APOSPC8593-500MG
{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl}methanol
{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl}methanol
Formel: C₁₂H₉F₄NOS
Molecular Weight: 291,27 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07369416
CAS-Nummer: 317319-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR