Methyl 5-chloro-6-oxo-1-[4-(trifluoromethyl)-benzyl]-1,6-dihydro-3-pyridinecarboxylate

Lieferant: Apollo Scientific

PC8594-10G PC8594-1G PC8594-5G
APOSPC8594-10GEA 477 EUR
APOSPC8594-10G APOSPC8594-1G APOSPC8594-5G
Methyl 5-chloro-6-oxo-1-[4-(trifluoromethyl)-benzyl]-1,6-dihydro-3-pyridinecarboxylate
Methyl-5-chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridinecarboxylate
Formel: C₁₅H₁₁ClF₃NO₃
Schmelzpunkt: 169…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027141
CAS-Nummer: 937602-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR