Methyl-4-(trifluormethyl)-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
PC8599-1G
APOSPC8599-1GEA
65.4
EUR
APOSPC8599-1G
Methyl-4-(trifluormethyl)-2-benzo[b]thiophencarboxylat
Methyl-4-(trifluormethyl)-2-benzo[b]thiophencarboxylat
Formel:
C₁₁H₇F₃O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027107 CAS-Nummer: 146137-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...