3-Bromo-2-fluorobenzoyl chloride 98%

Lieferant: Apollo Scientific
Gefahr

PC8604-1G
APOSPC8604-1GEA 58 EUR
APOSPC8604-1G
3-Bromo-2-fluorobenzoyl chloride 98%
3-Bromo-2-fluorobenzoyl chloride
Formel: C₇H₃BrClFO
Molecular Weight: 237,454 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864687
CAS-Nummer: 374554-41-3
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR