Methyl-7-(trifluormethyl)-2-benzo[b]thiophencarboxylat

Lieferant: Apollo Scientific
Achtung

PC8613-1G
APOSPC8613-1GEA 168 EUR
APOSPC8613-1G
Methyl-7-(trifluormethyl)-2-benzo[b]thiophencarboxylat
Methyl-7-(trifluormethyl)-2-benzo[b]thiophencarboxylat
Formel: C₁₁H₇F₃O₂S
Molecular Weight: 260.232 g/mol
Schmelzpunkt: 71…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027108
CAS-Nummer: 550998-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR