2-[4-(Trifluoromethoxy)phenyl]-1,3-thiazolane

Lieferant: Apollo Scientific

PC8614-10G PC8614-1G PC8614-5G
APOSPC8614-10GEA 568 EUR
APOSPC8614-10G APOSPC8614-1G APOSPC8614-5G
2-[4-(Trifluoromethoxy)phenyl]-1,3-thiazolane
2-[4-(Trifluoromethoxy)phenyl]-1,3-thiazolane
Formel: C₁₀H₁₀F₃NOS
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027148
CAS-Nummer: 937602-49-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR