2,4-Difluor-5-nitroanisol 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,5-Difluoro-2-methoxy-4-nitrobenzene
PC8621-5G
PC8621-1G
PC8621-25G
APOSPC8621-5GEA
139
EUR
APOSPC8621-5G
APOSPC8621-1G
APOSPC8621-25G
2,4-Difluor-5-nitroanisol 96%
2,4-Difluor-5-nitroanisol
Formel:
C₇H₅F₂NO₃ Molecular Weight: 189.1163 g/mol Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689675 CAS-Nummer: 179011-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...