4-[(2-Chlor-6-fluorbenzyl)oxy]-3-methoxybenzaldehyd 97%

Lieferant: Apollo Scientific

PC8635-1G
APOSPC8635-1GEA 69.6 EUR
APOSPC8635-1G
4-[(2-Chlor-6-fluorbenzyl)oxy]-3-methoxybenzaldehyd 97%
4-[(2-Chlor-6-fluorbenzyl)oxy]-3-methoxybenzaldehyd
Formel: C₁₅H₁₂ClFO₃
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01590672
CAS-Nummer: 306934-75-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR