3-[5-(Trifluoromethyl)-2-pyridinyl]aniline
Lieferant: Apollo Scientific
|
Achtung
|
PC8644-1G
PC8644-250MG
APOSPC8644-1GEA
434
EUR
APOSPC8644-1G
APOSPC8644-250MG
3-[5-(Trifluoromethyl)-2-pyridinyl]aniline
3-[5-(Trifluoromethyl)-2-pyridinyl]aniline
Formel:
C₁₂H₉F₃N₂ Schmelzpunkt: 84…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08276968 CAS-Nummer: 886361-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...