2-(4,4,5,5,6,6,6-Heptafluorohexyl)malonic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC8654-1G
APOSPC8654-1GEA
94.5
EUR
APOSPC8654-1G
2-(4,4,5,5,6,6,6-Heptafluorohexyl)malonic acid
2-(4,4,5,5,6,6,6-Heptafluorohexyl)malonic acid
Formel:
C₉H₉F₇O₄ Molecular Weight: 314,16 g/mol Schmelzpunkt: 114…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862051 CAS-Nummer: 244022-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...