2-[5-Fluoro-2-(pyridin-2-yl)-1H-indol-3-yl]ethylamine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-(2-Aminoethyl)-5-fluoro-2-(pyridin-2-yl)-1H-indole hydrochloride, 2-[3-(2-Aminoethyl)-5-fluoro-1H-indol-2-yl]pyridine hydrochloride

PC8681-250MG
APOSPC8681-250MGEA 764 EUR
APOSPC8681-250MG
2-[5-Fluoro-2-(pyridin-2-yl)-1H-indol-3-yl]ethylamine hydrochloride
2-[5-Fluoro-2-(pyridin-2-yl)-1H-indol-3-yl]ethylamine hydrochloride
Formel: C₁₅H₁₄FN₃·ClH
Molecular Weight: 291,751 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06245390
CAS-Nummer: 1049787-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR