3-(2-Aminoethyl)-5-fluoro-2-pyridin-3-yl-1H-indole hydrochloride

Lieferant: Apollo Scientific

Synonyme: 2-(5-Fluoro-2-pyridin-3-yl-1H-indol-3-yl)ethylamine hydrochloride

PC8682-500MG
APOSPC8682-500MGEA 315 EUR
APOSPC8682-500MG
3-(2-Aminoethyl)-5-fluoro-2-pyridin-3-yl-1H-indole hydrochloride
3-(2-Aminoethyl)-5-fluoro-2-pyridin-3-yl-1H-indole hydrochloride
Formel: C₁₅H₁₄FN₃·ClH
Molecular Weight: 291,751 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06245391
CAS-Nummer: 1049787-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR