5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Lieferant: Apollo Scientific
PC8683-1G
APOSPC8683-1GEA
575
EUR
APOSPC8683-1G
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Formel:
C₁₅H₁₀F₃N₃O₃ Molecular Weight: 337,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02253965 CAS-Nummer: 524036-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...