4-Chloro-2-(trifluoroacetyl)aniline hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: 1-(2-amino-5-chlorophenyl)-2,2,2-trifluoroethanone hydrochloride

PC8687-100G
APOSPC8687-100GEA 39.3 EUR
APOSPC8687-100G
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride 97%
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride
Formel: C₈H₆Cl₂F₃NO
Molecular Weight: 260,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263480
CAS-Nummer: 173676-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR