5-(Bromomethyl)-2-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC8704-250MG
PC8704-1G
PC8704-5G
APOSPC8704-250MGEA
0
EUR
APOSPC8704-250MG
APOSPC8704-1G
APOSPC8704-5G
5-(Bromomethyl)-2-(trifluoromethyl)pyridine
5-(Bromomethyl)-2-(trifluoromethyl)pyridine
Formel:
C₇H₅BrF₃N Schmelzpunkt: 37…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697684 CAS-Nummer: 108274-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...