4-(4-Methoxyphenyl)-5-[4-(trifluoromethoxy)phenyl]-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific

PC8731-1G
APOSPC8731-1GEA 0 EUR
APOSPC8731-1G
4-(4-Methoxyphenyl)-5-[4-(trifluoromethoxy)phenyl]-4H-1,2,4-triazole-3-thiol
4-(4-Methoxyphenyl)-5-[4-(trifluoromethoxy)phenyl]-4H-1,2,4-triazole-3-thiol
Formel: C₁₆H₁₂F₃N₃O₂S
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00098372
CAS-Nummer: 263707-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR