Ethyl-2-cyano-3-[4-(trifluoromethyl)phenyl]-acrylate
Lieferant: Apollo Scientific
|
Achtung
|
PC8740-1G
APOSPC8740-1GEA
140
EUR
APOSPC8740-1G
Ethyl-2-cyano-3-[4-(trifluoromethyl)phenyl]-acrylate
Ethyl-2-cyano-3-[4-(trifluoromethyl)phenyl]-acrylate
Formel:
C₁₃H₁₀F₃NO₂ Molecular Weight: 269,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00176452 CAS-Nummer: 149550-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...