Ethyl-2-cyano-3-[3-(trifluoromethyl)-phenyl]acrylate
Lieferant: Apollo Scientific
|
Achtung
|
PC8741-1G
APOSPC8741-1GEA
139
EUR
APOSPC8741-1G
Ethyl-2-cyano-3-[3-(trifluoromethyl)-phenyl]acrylate
Ethyl-2-cyano-3-[3-(trifluoromethyl)-phenyl]acrylate
Formel:
C₁₃H₁₀F₃NO₂ Schmelzpunkt: 77…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03283527 CAS-Nummer: 149550-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...