4-Fluor-2-(hydroxymethyl)benzonitril
Lieferant: Apollo Scientific
|
Achtung
|
PC8753-1G
PC8753-5G
APOSPC8753-1GEA
105
EUR
APOSPC8753-1G
APOSPC8753-5G
4-Fluor-2-(hydroxymethyl)benzonitril
4-Fluor-2-(hydroxymethyl)benzonitril
Formel:
C₈H₆FNO Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864693 CAS-Nummer: 1000339-93-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...