2-Methoxy-5-(trifluormethoxy)benzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC8760-10G
PC8760-5G
PC8760-1G
APOSPC8760-10GEA
203
EUR
APOSPC8760-10G
APOSPC8760-5G
APOSPC8760-1G
2-Methoxy-5-(trifluormethoxy)benzoesäure 98%
2-Methoxy-5-(trifluoromethoxy)benzoic acid
Formel:
C₉H₇F₃O₄ Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862055 CAS-Nummer: 191604-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...