4-Nitrophenyl trifluoromethyl sulphoxide
Lieferant: Apollo Scientific
Synonyme:
1-Nitro-4-[(trifluoromethyl)sulphinyl]benzene
PC8765-500MG
APOSPC8765-500MGEA
865
EUR
APOSPC8765-500MG
4-Nitrophenyl trifluoromethyl sulphoxide
4-Nitrophenyl trifluoromethyl sulphoxide
Formel:
C₇H₄F₃NO₃S Molecular Weight: 239,172 g/mol Schmelzpunkt: 59…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00656389 CAS-Nummer: 394-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...