3-Chloro-4-trifluoromethoxyphenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC8774-5G
APOSPC8774-5GEA
140
EUR
APOSPC8774-5G
3-Chloro-4-trifluoromethoxyphenol 98%
3-Chloro-4-trifluoromethoxyphenol
Formel:
C7H4ClF3O2 Molecular Weight: 212.5537 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972754 CAS-Nummer: 1000339-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...