4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-butyn-2-ol

Lieferant: Apollo Scientific

PC8781-500MG
APOSPC8781-500MGEA 560 EUR
APOSPC8781-500MG
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-butyn-2-ol
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-butyn-2-ol
Formel: C₁₀H₇ClF₃NO
Siedepunkt: 155 °C/0,4mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD08443949
CAS-Nummer: 937601-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR