4-(4-Fluorobenzoyl)-1H-pyrrole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC8782-1G
APOSPC8782-1GEA 336 EUR
APOSPC8782-1G
4-(4-Fluorobenzoyl)-1H-pyrrole-2-carboxylic acid
4-(4-Fluorobenzoyl)-1H-pyrrole-2-carboxylic acid
Formel: C₁₂H₈FNO₃
Schmelzpunkt: 223…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443967
CAS-Nummer: 924868-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR