2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid

Lieferant: Apollo Scientific
Achtung

PC8786-250MG
APOSPC8786-250MGEA 77 EUR
APOSPC8786-250MG
2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
Formel: C₁₃H₁₀F₃NO₂S
Molecular Weight: 301,29 g/mol
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443969
CAS-Nummer: 924868-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR