6-Propyl-4-(trifluoromethyl)-2-pyridinol

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-6-(prop-1-yl)-4-(trifluoromethyl)pyridine

PC8787-1G
APOSPC8787-1GEA 538 EUR
APOSPC8787-1G
6-Propyl-4-(trifluoromethyl)-2-pyridinol
6-Propyl-4-(trifluoromethyl)-2-pyridinol
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,18 g/mol
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443946
CAS-Nummer: 937601-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR