tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-(4-amino-2-fluorophenyl)piperazine-1-carboxylate, 4-(4-Amino-2-fluorophenyl)piperazine, N1-BOC protected

PC8788-250MG
APOSPC8788-250MGEA 21.9 EUR
APOSPC8788-250MG
tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Formel: C₁₅H₂₂FN₃O₂
Molecular Weight: 295,36 g/mol
Schmelzpunkt: 110…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08443960
CAS-Nummer: 154590-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR