2-Bromo-4'-(trifluoromethyl)propiophenone

Lieferant: Apollo Scientific
Gefahr

PC8809-1G PC8809-250MG PC8809-5G
APOSPC8809-1GEA 201 EUR
APOSPC8809-1G APOSPC8809-250MG APOSPC8809-5G
2-Bromo-4'-(trifluoromethyl)propiophenone
2-Bromo-4'-(trifluoromethyl)propiophenone
Formel: C₁₀H₈BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD17010234
CAS-Nummer: 95728-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR