2-Bromo-6-methyl-4-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC8816-1G
PC8816-250MG
APOSPC8816-1GEA
179
EUR
APOSPC8816-1G
APOSPC8816-250MG
2-Bromo-6-methyl-4-(trifluoromethyl)pyridine
2-Bromo-6-methyl-4-(trifluoromethyl)pyridine
Formel:
C₇H₅BrF₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368051 CAS-Nummer: 451459-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...