2-Bromo-6-methyl-4-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC8816-1G PC8816-250MG
APOSPC8816-1GEA 179 EUR
APOSPC8816-1G APOSPC8816-250MG
2-Bromo-6-methyl-4-(trifluoromethyl)pyridine
2-Bromo-6-methyl-4-(trifluoromethyl)pyridine
Formel: C₇H₅BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD07368051
CAS-Nummer: 451459-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR