3-Chlor-5-(trifluormethyl)picolinonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-5-(trifluoromethyl)pyridine-2-carbonitrile
PC8824-1G
PC8824-5G
PC8824-10G
APOSPC8824-1GEA
21.9
EUR
APOSPC8824-1G
APOSPC8824-5G
APOSPC8824-10G
3-Chlor-5-(trifluormethyl)picolinonitril
3-Chlor-5-(trifluormethyl)picolinonitril
Formel:
C₇H₂ClF₃N₂ Molecular Weight: 206.5524 g/mol Siedepunkt: 90 °C/11mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD00664066 CAS-Nummer: 80194-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...