3-Chlor-5-(trifluormethyl)picolinonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-5-(trifluoromethyl)pyridine-2-carbonitrile

PC8824-1G PC8824-5G PC8824-10G
APOSPC8824-1GEA 21.9 EUR
APOSPC8824-1G APOSPC8824-5G APOSPC8824-10G
3-Chlor-5-(trifluormethyl)picolinonitril
3-Chlor-5-(trifluormethyl)picolinonitril
Formel: C₇H₂ClF₃N₂
Molecular Weight: 206.5524 g/mol
Siedepunkt: 90 °C/11mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD00664066
CAS-Nummer: 80194-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR