1-(4-(4-Amino-2-(trifluoromethyl)phenyl)piperazin-1-yl)ethanone
Lieferant: Apollo Scientific
|
Achtung
|
PC8825-1G
APOSPC8825-1GEA
125
EUR
APOSPC8825-1G
1-(4-(4-Amino-2-(trifluoromethyl)phenyl)piperazin-1-yl)ethanone
1-(4-(4-Amino-2-(trifluoromethyl)phenyl)piperazin-1-yl)ethanone
Formel:
C₁₃H₁₆F₃N₃O Lagertemperatur: Kühlschrank |
CAS-Nummer:
914348-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...