2-Amino-4-fluorpyridin
Lieferant: Apollo Scientific
PC8826-1G
APOSPC8826-1GEA
21.9
EUR
APOSPC8826-1G
2-Amino-4-fluorpyridin
2-Amino-4-fluoropyridine
Formel:
C₅H₅FN₂ Molecular Weight: 112,04 g/mol |
MDL:
MFCD09260906 CAS-Nummer: 944401-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...