2-Brom-3-fluorpyridin 98%

Lieferant: Apollo Scientific
Achtung

PC8846-5G PC8846-100G PC8846-25G
APOSPC8846-5GEA 21.9 EUR
APOSPC8846-5G APOSPC8846-100G APOSPC8846-25G
2-Brom-3-fluorpyridin 98%
2-Bromo-3-fluoropyridine
Formel: C₅H₃BrFN
Molecular Weight: 175.99 g/mol
Dichte: 1.74 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD04114128
CAS-Nummer: 40273-45-8
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR