2-Fluoro-4-(phenylsulphonyl)phenylhydrazine
Lieferant: Apollo Scientific
PC8858-1G
PC8858-5G
APOSPC8858-1GEA
166
EUR
APOSPC8858-1G
APOSPC8858-5G
2-Fluoro-4-(phenylsulphonyl)phenylhydrazine
2-Fluoro-4-(phenylsulphonyl)phenylhydrazine
Formel:
C₁₂H₁₁FN₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800751 CAS-Nummer: 1000339-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...