1-[6-Fluoro-2-(trifluoromethyl)quinolin-4-yl]piperazine 97%

Lieferant: Apollo Scientific
Achtung

PC8864-100MG
APOSPC8864-100MGEA 58 EUR
APOSPC8864-100MG
1-[6-Fluoro-2-(trifluoromethyl)quinolin-4-yl]piperazine 97%
1-[6-Fluoro-2-(trifluoromethyl)quinolin-4-yl]piperazine
Formel: C₁₄H₁₃F₄N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03094523
CAS-Nummer: 541539-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR