1-[6-Chloro-2-(trifluoromethyl)quinol-4-yl]piperazine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8865-250MG
APOSPC8865-250MGEA
91.5
EUR
APOSPC8865-250MG
1-[6-Chloro-2-(trifluoromethyl)quinol-4-yl]piperazine 97%
1-[6-Chloro-2-(trifluoromethyl)quinol-4-yl]piperazine
Formel:
C₁₄H₁₃ClF₃N₃ Molecular Weight: 315.721 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094524 CAS-Nummer: 401567-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...