N,N-Dimethyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide

Lieferant: Apollo Scientific

Synonyme: 2-[4-(N,N-Dimethylsulphamoyl)phenoxy]-4-(trifluoromethyl)pyridine

PC8874-1G
APOSPC8874-1GEA 361 EUR
APOSPC8874-1G
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Formel: C₁₄H₁₃F₃N₂O₃S
Molecular Weight: 346,325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109524
CAS-Nummer: 1257535-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR