N,N-Dimethyl-4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide

Lieferant: Apollo Scientific

Synonyme: 2-[4-(N,N-Dimethylsulphamoyl)phenoxy]-5-(trifluoromethyl)pyridine

PC8875-1G
APOSPC8875-1GEA 302 EUR
APOSPC8875-1G
N,N-Dimethyl-4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
N,N-Dimethyl-4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Formel: C₁₄H₁₃F₃N₂O₃S
Molecular Weight: 346.325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109525
CAS-Nummer: 1227954-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR