N,N-Dimethyl-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide

Lieferant: Apollo Scientific

Synonyme: 2-[4-(N,N-Dimethylsulphamoyl)phenoxy]-6-(trifluoromethyl)pyridine

PC8876-1G
APOSPC8876-1GEA 310 EUR
APOSPC8876-1G
N,N-Dimethyl-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
N,N-Dimethyl-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Formel: C₁₄H₁₃F₃N₂O₃S
Molecular Weight: 346,325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109526
CAS-Nummer: 1227954-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR