N-Methyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Lieferant: Apollo Scientific
Synonyme:
2-[4-(N-Methylsulphamoyl)phenoxy]-4-(trifluoromethyl)pyridine
PC8878-1G
APOSPC8878-1GEA
361
EUR
APOSPC8878-1G
N-Methyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
N-Methyl-4-{[4-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Formel:
C₁₃H₁₁F₃N₂O₃S Molecular Weight: 332,298 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109528 CAS-Nummer: 1257535-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...